Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 6
Positive control: N2A, SW620, rat uterus tissue, human placenta tissue, mouse brain tissue.
Predicted band size: 48 kDa
Subcellular Location: Membrane.
The KIR2 subunit family includes 2. 1, 2. 2, 2. 3 and 2. 4. Unlike G protein-coupled KIR3 subunits, KIR2. 1 requires both phosphorylation by PKA and ATP hydrolysis for functional activity. KIR2. 1 is expressed in the superior and inferior collicula and the pontine region of the brain, where it moderates synaptic transmission, like many other potassium channels. In the placenta, KIR2. 1 is expressed throughout gestation in cytotrophoblast cells. In the kidney, KIR2. 1 colocalizes with KIR5. 1 in the proximal tubule. KIR2. 1, 2. 2 and 2. 3 associate with the membrane-associated guanylate kinase synapseassociated protein 97 in the cerebellum and heart. Phosphorylation of KIR2. 2 by protein kinase A inhibits the associates with SAP97.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cardiac inward rectifier potassium channel; HHIRK 1; HIRK 1; hIRK1; Inward rectifier K; Inward rectifier K(+) channel Kir2.1; inward rectifier K+ channel KIR2.1; Inward rectifier potassium channel 2; IRK 1; IRK-1; IRK2; LQT 7; Potassium channel, inwardly rectifying subfamily J member 2; potassium channel, inwardly rectifying subfamily J, member 2; potassium inwardly-rectifying channel, subfamily J, member 2; RBL-IRK1; SQT 3
Gene Aliases: ATFB9; HHBIRK1; HHIRK1; IRK1; Kcnf1; KCNJ2; KIR2.1; LQT7; SQT3
UniProt ID: (Human) P63252, (Rat) Q64273, (Mouse) P35561
Entrez Gene ID: (Human) 3759, (Rat) 29712, (Mouse) 16518
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support