Invitrogen
MA1-70114 detects Kudoa Thrysites in Kudoa thrysites samples.
MA1-70114 has been successfully used in Western blot, immunofluorescence and flow cytometry procedures.
The MA1-70114 immunogen is PercollTM-purified K. thyrsites spores.
Kudoa thrysites is a myxosporean parasite of marine fishes. It has a worldwide distribution, and infects a wide range of host species. This parasite is responsible for causing economic losses to the fisheries sector, by causing post-mortem "myoliquefaction", a softening of the flesh to such an extent that the fish becomes unmarketable. It is not infective to humans.
Protein Aliases: myxosporean