Search Thermo Fisher Scientific
Nordic
Reconstituted by adding 0.5 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20°C.
Cross-reactivities against enzymes of other sources may occur but have not been determined.
Prior to use, an aliquot is thawed slowly at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4°C, not refrozen, and preferably used t he same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the product.
L-amino-acid oxidase catalyzes an oxidative deamination of predominantly hydrophobic and aromatic L-amino acids, thus producing hydrogen peroxide that may contribute to the diverse toxic effects of this enzyme. Shows activity on L-Leu. Exhibits diverse biological activities, such as hemorrhage, hemolysis, edema, apoptosis of vascular endothelial cells or tumor cell lines, and antiparasitic activities, as well as regulation of platelet aggregation. Effects of snake L-amino oxidases on platelets are controversial, since they either induce aggregation or inhibit agonist-induced aggregation. These different effects are probably due to different experimental conditions. In addition, this protein inhibits dose-dependently the growth of Gram-positive, Gram-negative bacteria and yeast, probably by the generation of hydrogen peroxide.