Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVPALRYLV GACGRARGRF AGGSPGACGF ASGRPRPLCG GSRSASTSSF DIVIVGGGIV GLASARALIL RHPSLSIGVL EKEKDLAVHQ TGHNSGVIHS GIYYKPESLK AKLCVQGAAL LYEYCQQKGI SYKQCGKLIV AVEQEEIPRL QALYEKGLQN GVPGLRLIQQ EDIKKKEPYC RGLMAIDCPH TGIVDYRQVA LSFAQDFQEA GGSVLTNFEV KGIEMAKESP SRSIDGMQYP IVIKNTKGEE IRCQYVVTCA GLYSDRISEL SGCTPDPRIV PFRGDYLLLK PEKCYLVKGN IYPVPDSRFP FLGVHFTPRM DGSIWLGPNA VLAFKREGYR PFDFSATDVM DI (1-351 aa encoded by BC006117)
This gene encodes L-2-hydroxyglutarate dehydrogenase, a FAD-dependent enzyme that oxidizes L-2-hydroxyglutarate to alpha-ketoglutarate in a variety of mammalian tissues. Mutations in this gene cause L-2-hydroxyglutaric aciduria, a rare autosomal recessive neurometabolic disorder resulting in moderate to severe mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-hydroxyglutarate dehydrogenase; alpha-hydroxyglutarate oxidoreductase; alpha-ketoglutarate reductase; Duranin; FLJ12618; L-2-hydroxyglutarate dehydrogenase, mitochondrial; L-alpha-hydroxyglutarate dehydrogenase
Gene Aliases: C14orf160; L2HGA; L2HGDH
UniProt ID: (Human) Q9H9P8
Entrez Gene ID: (Human) 79944, (Pig) 100152180
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support