Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes L-2-hydroxyglutarate dehydrogenase, a FAD-dependent enzyme that oxidizes L-2-hydroxyglutarate to alpha-ketoglutarate in a variety of mammalian tissues. Mutations in this gene cause L-2-hydroxyglutaric aciduria, a rare autosomal recessive neurometabolic disorder resulting in moderate to severe mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-hydroxyglutarate dehydrogenase; alpha-hydroxyglutarate oxidoreductase; alpha-ketoglutarate reductase; Duranin; FLJ12618; L-2-hydroxyglutarate dehydrogenase, mitochondrial; L-alpha-hydroxyglutarate dehydrogenase
Gene Aliases: C14orf160; L2HGA; L2HGDH
UniProt ID: (Human) Q9H9P8
Entrez Gene ID: (Human) 79944, (Pig) 100152180
Molecular Function: dehydrogenase oxidoreductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support