Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRRREGHGTD SEMGQGPVRE SQSSDPPALQ FRISEYKPLN MAGVEQPPTP ELRQEGVTEY EDGGAPAGDG EAGPQQAEDH PQNPPEDPNQ DPPEDDSTCQ CQACGPHQAA GPDLGSSNDG CPQLFQERSV IVENSSGSTS ASELLKPMKK RKRREYQSPS EEESEPEAME KQEEGKDPEG QPTASTPESE EWSSSQPATG EKKECWSWES YLEEQKAITA PVSLFQDSQA VTHNKNGFKL GMKLEGIDPQ HPSMYFILTV AEVCGYRLRL HFDGYSECHD FWVNANSPDI HPAGWFEKTG HKLQPPKGYK EEEFSWSQYL RSTRAQAAPK HLFVSQSHSP PPLGFQVGMK LEAVDRMNPS LVCVASVTDV VDSRFLVHFD NWDDTYDYWC DPSSPYIHPV GWCQKQGKPL TPPQDYPDPD NFCWEKYLEE TGASAVPTWA FKVRPPHSFL VNMKLEAVDR RNPALIRVAS VEDVEDHRIK IHFDGWSHGY DFWIDADHPD IHPAGWCSKT GHPLQPPLGP REPSSASPGG CPPLSYRSLP HTRTSKYSFH HRKCPTPGCD GSGHVTGKFT AHHCLSGCPL AERNQSRLKA ELSDSEASAR KKNLSGFSPR KKPRHHGRIG RPPKYRKIPQ EDFQTLTPDV VHQSLFMSAL SAHPDRSLSV CWEQHCKLLP GVAGISASTV AKWTIDEVFG FVQTLTGCED QARLFKDEAR IVRVTHVSGK TLVWTVAQLG DLVCSDHLQE GKGILETGVH SLLCSLPTHL LAKLSFASDS QY
L3MBTL1 is the homolog of a protein identified in Drosophila as a suppressor of malignant transformation of neuroblasts and ganglion-mother cells in the optic centers of the brain. L3MBTL1 is localized to condensed chromosomes in mitotic cells and overexpression of this protein in a glioma cell line results in improper nuclear segregation and cytokinesis producing multinucleated cells. L3MBTL1 represents a polycomb group gene the encoded protein functions to regulate gene activity, likely via chromatin modification. Polycomb group protein specifically recognizes and binds mono- and dimethyllysine residues on target proteins, thereby acting as a reader of a network of post-translational modifications. Alternatively spliced transcript variants encoding different isoforms of the L3MBTL1 gene have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-l(3)mbt; L(3)mbt protein homolog; L3MBTL1; lethal (3) malignant brain tumor l(3); Lethal(3)malignant brain tumor-like protein 1; RP1-138B7.1
Gene Aliases: dJ138B7.3; H-L(3)MBT; KIAA0681; L3MBT; L3MBTL; L3MBTL1; ZC2HC3
UniProt ID: (Human) Q9Y468
Entrez Gene ID: (Human) 26013
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support