Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-l(3)mbt-like protein 3; L(3)mbt-like protein 3; Lethal(3)malignant brain tumor-like protein 3; MBT-1
Gene Aliases: AI481284; KIAA1798; L3MBTL3; MBT-1; MBT1
UniProt ID: (Human) Q96JM7, (Mouse) Q8BLB7
Entrez Gene ID: (Human) 84456, (Mouse) 237339, (Rat) 309550
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support