Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKQPNRKRK LNMDSKERLD QDGRLEQAEE EKKPKDSTTP LSHVPSAAAQ GAWSWEWYLK EQKAVAAPVE LFSKDQSFPE HENGFQIGMR LE (1-91 aa encoded by BC039316)
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-l(3)mbt-like protein 4; L(3)mbt-like protein 4; Lethal(3)malignant brain tumor-like protein 4
Gene Aliases: A730037L19Rik; D930040M24Rik; HsT1031; L3MBTL4
UniProt ID: (Human) Q8NA19, (Mouse) B1B1A0
Entrez Gene ID: (Human) 91133, (Mouse) 320858
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support