Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MKQPNRKRKL NMDSKERLDQ DGRLEQAEEE KKPKDSTTPL SHVPSAAAQG AWSWEWYLKE QKAVAAPVEL FSKDQSFPEH ENGFQIGMRL EGIDPRHPSV FCVLSVAEVC GYRLRLHFDG YLSCYDFWTN AGSPDIHPVG WCEKTKHELH IPKGYRKDKF VWMDYLKACK LQNAPKKLFR NRSPNGPMSK EFQVGMKLEA VDRKNPSLVC VATIADIVED RLLVHFDNWD DSYDYWCDVN SPYVQPVGWC QENGRTLIAP QGYPNPENFS WTEYLEATQT NAVPAKVFKM RLPHGFLPNM KLEVVDKRNP RLIRVATIVD VDDQRVKVHF DGWDHKYDYW VEADSPDIHP IGWCDVTGHP LEVPQRTNDL KILPGQAVCP TPGCRGIGHI RGPRYSGHHS AFGCPYSDMN LKKEATLHDR LREQTQANLE SDSSHSKSKS LCSLNFNGKH EKVNSQPRLV QQAKCLKIKG KEDIDLDNLF RGRVRWLMPV IPALWEAEAG RSRDQEIKTI LANAVKPRLY QKYKKLARRG GGCL
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-l(3)mbt-like protein 4; l(3)mbt-like protein 4; Lethal(3)malignant brain tumor-like protein 4
Gene Aliases: HsT1031; L3MBTL4
UniProt ID: (Human) Q8NA19
Entrez Gene ID: (Human) 91133
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support