Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: NSNENLLPGY LKPETMVMMW TPVPNTEMSW DKEGKYAMAW GVVEKKQTYG SCRKQRHYAS HTGGAVGASS VLLVLPEELD TETINNKVP
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.
This gene encodes a protein from the large 39S subunit of the mitochondrial ribosome (mitoribosome). The encoded protein has some sequence similarity to prokaryotic beta-lactamases but most of the residues that are responsible for the beta-lactamase activity are not conserved between the two proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochondrial 39S ribosomal protein L56; Serine beta-lactamase-like protein LACTB, mitochondrial
Gene Aliases: G24; LACTB; MRPL56; UNQ843/PRO1781
UniProt ID: (Human) P83111
Entrez Gene ID: (Human) 114294
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support