Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAAVLQRVER LSNRVVRVLG CNPGPMTLQG TNTYLVGTGP RRILIDTGEP AIPEYISCLK QALTEFNTAI QEIVVTHWHR DHSGGIGDIC KSINNDTTYC IKKLPRNPQR EEIIGNGEQQ YVYLKDGDVI KTEGATLRVL YTPGHTDDHM ALLLEEENAI FSGDCILGEG TTVFEDLYDY MNSLKELLKI KADIIYPGHG PVIHNAEAKI QQYISHRNIR EQQILTLFRE NFEKSFTVME LVKIIYKNTP ENLHEMAKHN LLLHLKKLEK EGKIFSNTDP DKKWKAHL
Endoribonuclease; cleaves preferentially 3' to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3' -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-lactamase-like protein 2; Endoribonuclease LACTB2; lactamase, beta 2; testicular secretory protein Li 23
Gene Aliases: CGI-83; LACTB2
UniProt ID: (Human) Q53H82
Entrez Gene ID: (Human) 51110
Molecular Function:
endoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support