Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
HOPX is an atypical homeodomain protein which does not bind DNA and is required to modulate cardiac growth and development. HOPX acts via its interaction with SRF, thereby modulating the expression of SRF-dependent cardiac-specific genes and cardiac development. HOPX prevents SRF-dependent transcription either by inhibiting SRF binding to DNA or by recruiting histone deacetylase (HDAC) proteins that prevent transcription by SRF. HOPX overexpression causes cardiac hypertrophy and may act as a tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GIIg15b; global ischemia induced protein GIIG15B; Global ischemia-induced gene 15B protein; global ischemia-induced protein 15B; homeobox only domain; Homeobox-only protein; homeodomain only protein; Homeodomain-only protein; Lung cancer-associated Y protein; mOB1; not expressed in choriocarcinoma clone 1; Not expressed in choriocarcinoma protein 1; odd homeobox 1; Odd homeobox protein 1
Gene Aliases: 1110018K11Rik; 1200015P04Rik; 2300002F06Rik; AI848177; AW490897; CAMEO; GIIg15b; Hdop; HOD; HOP; HOPX; LAGY; NECC1; OB1; Obl; SMAP31; TOTO
UniProt ID: (Human) Q9BPY8, (Mouse) Q8R1H0, (Rat) Q78ZR5
Entrez Gene ID: (Human) 84525, (Mouse) 74318, (Rat) 171160
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support