Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MSPHPTALLG LVLCLAQTIH TQEEDLPRPS ISAEPGTVIP LGSHVTFVCR GPVGVQTFRL ERESRSTYND TEDVSQASPS ESEARFRIDS VSEGNAGPYR CIYYKPPKWS EQSDYLELLV KETSGGPDSP DTEPGSSAGP TQRPSDNSHN EHAPASQGLK AEHLYILIGV SVVFLFCLLL LVLFCLHRQN QIKQGPPRSK DEEQKPQQRP DLAVDVLERT ADKATVNGLP EKDRETDTSA LAAGSSQEVT YAQLDHWALT QRTARAVSPQ STKPMAESIT YAAVARH
LAIR1 (CD305) functions as an inhibitory receptor that plays a constitutive negative regulatory role on cytolytic function of natural killer (NK) cells, B-cells and T-cells. Activation by Tyr phosphorylation results in recruitment and activation of the phosphatases PTPN6 and PTPN11. It also reduces the increase of intracellular calcium evoked by B-cell receptor ligation. May also play its inhibitory role independently of SH2-containing phosphatases. Modulates cytokine production in CD4+ T-cells, down-regulating IL2 and IFNG production while inducing secretion of transforming growth factor beta. Down-regulates also IgG and IgE production in B-cells as well as IL8, IL10 and TNF secretion. Inhibits proliferation and induces apoptosis in myeloid leukemia cell lines as well as prevents nuclear translocation of NF-kappa-B p65 subunit/RELA and phosphorylation of I-kappa-B alpha/CHUK in these cells. Inhibits the differentiation of peripheral blood precursors towards dendritic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD305; immunoglobulin heavy chain variable region; LAIR-1; leukocyte-associated Ig-like receptor 1; Leukocyte-associated immunoglobulin-like receptor 1
Gene Aliases: 5133400O11Rik; BB115266; CD305; D7Bwg0421e; LAIR-1; LAIR1
UniProt ID: (Human) Q6GTX8, (Mouse) Q8BG84
Entrez Gene ID: (Human) 3903, (Mouse) 52855
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support