Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs and Western blots, no cross-reactivity with recombinant human LAIR1, recombinant mouse (rm) LAIR1 or rmLAIR2 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the immunoglobulin superfamily. It was identified by its similarity to leukocyte-associated immunoglobulin-like receptor 1, a membrane-bound receptor that modulates innate immune response. The protein encoded by this locus is a soluble receptor that may play roles in both inhibition of collagen-induced platelet aggregation and vessel formation during placental implantation. This gene maps to a region of 19q13.4, termed the leukocyte receptor cluster, which contains 29 genes in the immunoglobulin superfamily. Alternatively spliced transcript variants have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD306; CD306 antigen; LAIR-2; leukocyte-associated Ig-like receptor 2; leukocyte-associated Ig-like receptor-2; Leukocyte-associated immunoglobulin-like receptor 2; MGC71634
Gene Aliases: CD306; LAIR2
UniProt ID: (Human) Q6ISS4
Entrez Gene ID: (Human) 3904
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support