Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Immunogen sequence: LELNLTDSEN ATCLYAKWQM NFTVRYETTN KTYKTVTISD HGTVTYNGSI CGDDQNGPKI AVQFGPGFSW IANFTKAAST YSIDSVSFSY NTGDNTTFPD AEDKGILTVD ELLAIRIPLN DLFRCNSLST LEKNDVVQHY WDVLVQAFVQ NGTVSTNEFL CDKDKTSTVA PTIHTTVPSP TTTPTPKEKP EAGTYSVNNG NDTCLLATMG LQLNITQDKV ASVININPNT THSTGSCRSH TALLRLNSST IKYLDFVFAV KNENRFYLKE VNISMYLVNG SVFSIANNNL SYWDAPLGSS YMCNKEQTVS VSGAFQINTF DLRVQPFNVT QGKYSTAQDC SADDDNF
LAMP2 (Lysosome-associated membrane glycoprotein 2, CD107b) is a member of a family of membrane glycoproteins. LAMP2 provides selectins with carbohydrate ligands, and may play a role in tumor cell metastasis, and have a function in the protection, maintenance, and adhesion of the lysosome. Prior to posttranslational modification, LAMP2 is approximately 46 kDa polypeptide. Mature, functional LAMP2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of about 100-130 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. LAMP2 is used to monitor a very selective form of autophagy called chaperone-mediated autophagy. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. The LAMP protein has 3 variant forms: LAMP-2A, -2B, and 2C. LAMP2, together with CD107a/LAMP-1, is a major constituent of lysosomal membrane. The LAMP proteins are involved in lysosome biogenesis and are required for fusion of lysosomes with phagosomes, and LAMP2 is an important regulator in successful phagosomal maturation. LAMP2 deficiency causes an accumulation of autophagosomes in many tissues leading to cardiomyopathy and myopathy (Danon’s disease).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD107 antigen-like family member B; CD107b; Igp96; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; LGP-96; lysosomal-associated membrane protein 2; Lysosome-associated membrane glycoprotein 2; RP23-193O17.2
Gene Aliases: CD107b; LAMP-2; LAMP2; LAMPB; LGP110
UniProt ID: (Human) P13473
Entrez Gene ID: (Human) 3920
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support