Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: TKRSITFLCG DAGPLAVAAV LYHKMNNEKQ AEDCITRLIH LNKIDPHAPN EMLYGRIGYI YALLFVNKNF GVEKIPQSHI QQICETILTS GENLARKRNF TAKSPLMYEW YQEYYVGAAH GLAGIYYYLM QPSLQVSQGK LHSLVKPSVD YVCQLKFPSG NYPPCIGDNR DLLVHWCHGA PGVIYMLIQA YKVFREEKYL CDAYQCADVI WQYGLLKKGY GLCHGSAGNA YAFLTLYNLT QDMKYLYRAC KFAEWCLEYG EHGCRTPDTP FSLFEGMAGT IYFLADLLVP TKARFPAFEL
NRIP is a novel 860a.a nuclear protein consisting of seven conserved WD40 domains and one NLS motif. It binds to androgen and glucocorticoid receptors and up regulates their transcriptional activity, thereby functioning as a nuclear receptor coactivator. Role of NRIP has been implicated in cell growth and also in cervical and prostrate cancer thus indicating a therapeutic intervention. Northern Blot analysis detected a high expression of NRIP in skeletal muscle and testis and low expression in heart, prostrate and adrenal gland.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40 kDa erythrocyte membrane protein; G protein-coupled receptor 69; G protein-coupled receptor 69A; Glutathione S-transferase LANCL1; LanC (bacterial lantibiotic synthetase component C)-like 1; LanC (bacterial lantibiotic synthetase component); LanC lantibiotic synthetase component C-like 1; LanC-like 1; LanC-like 1(bacterial); LanC-like protein 1; p40
Gene Aliases: AW124738; Gpr69; GPR69A; LANCL1; p40
UniProt ID: (Human) O43813, (Rat) Q9QX69, (Mouse) O89112
Entrez Gene ID: (Pig) 100153701, (Human) 10314, (Rat) 114515, (Mouse) 14768
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support