Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
LARP4 gene ontology annotations related to this gene include RNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Mpl binding protein; La ribonucleoprotein domain family member 4; La ribonucleoprotein domain family, member 4; La-related protein 4
Gene Aliases: DXErtd793; DXErtd793e; LARP4; PP13296
UniProt ID: (Human) Q71RC2, (Mouse) Q8BWW4
Entrez Gene ID: (Human) 113251, (Mouse) 207214
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support