Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLLFVEQVAS KGTGLNPNAK VWQEIAPGNT DATPVTHGTE SSWHEIAATS GAHPEVSVFN TGNAELSEDI CKEYEVMYSS SCETTRNTTG IEESTDGMIL GPEDLSYQIY DVSGESNSAV STEDLKECLK KQLEFCFSRE NLSKDLYLIS QMDSDQFIPI WTVANMEEIK KLTTDPDLIL EVLRSSPMVQ VDEKGEKVRP SHKRCIVILR EIPETTPIEE VKGLFKSENC PKVISCEFAH NSNWYITFQS DTDAQQAFKY LREEVKTFQG KPIMARIKAI NTFFAKNGYR LMDSSIYSHP IQTQAQYASP VFMQPVYNPH QQYSVYSIVP QSWSPNPTPY FETPLAPFPN GSFVNGFNSP GSYKTNAAAM NMGRPFQKNR VKPQFRSSGG SEHSTEGSVS LGDGQLNRYS SRNFPAERHN PTVTGHQEQT YLQKETSTLQ VEQNGDYGRG RRTLFRGRRR REDDRISRPH PSTAESKAPT PKFDLLASNF PPLPGSSSRM PGELVLENRM SDVVKGVYKE KDNEELTISC PVPADEQTEC TSAQQLNMST SSPCAAELTA LSTTQQEKDL IEDSSVQKDG LNQTTIPVSP PSTTKPSRAS TASPCNNNIN AATAVALQEP RKLSYAEVCQ KPPKEPSSVL VQPLRELRSN VVSPTKNEDN GAPENSVEKP HEKPEARASK DYSGFRGNII PRGAAGKIRE QRRQFSHRAI PQGVTRRNGK EQYVPPRSPK
LARP4 gene ontology annotations related to this gene include RNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Mpl binding protein; La ribonucleoprotein domain family member 4; La ribonucleoprotein domain family, member 4; La-related protein 4; la-related protein 4-like
Gene Aliases: LARP4; PP13296
UniProt ID: (Human) Q71RC2
Entrez Gene ID: (Human) 113251, (Rat) 100910299
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support