Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
LARP7 gene ontology annotations related to this gene include RNA processing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP564K112; hLARP7; La ribonucleoprotein domain family member 7; La ribonucleoprotein domain family, member 7; La-related protein 7; P-TEFb-interaction protein for 7SK stability; PIP7S
Gene Aliases: ALAZS; C330027G06Rik; D3Wsu161e; HDCMA18P; LARP7; PIP7S
UniProt ID: (Human) Q4G0J3, (Mouse) Q05CL8
Entrez Gene ID: (Human) 51574, (Mouse) 28036
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support