Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
This gene encodes a cytosolic leucine-tRNA synthetase, a member of the class I aminoacyl-tRNA synthetase family. The encoded enzyme catalyzes the ATP-dependent ligation of L-leucine to tRNA(Leu). It is found in the cytoplasm as part of a multisynthetase complex and interacts with the arginine tRNA synthetase through its C-terminal domain. Alternatively spliced transcript variants of this gene have been found; however, their full-length nature is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoplasmic leucyl-tRNA synthetase; cytosolic leucyl-tRNA synthetase; leucine translase; leucine tRNA ligase 1, cytoplasmic; Leucine--tRNA ligase, cytoplasmic; leucine-tRNA ligase; Leucyl-tRNA synthetase; leucyl-tRNA synthetase, cytoplasmic; LeuRS; proliferation-inducing gene 44
Gene Aliases: 2310045K21Rik; 3110009L02Rik; AW536573; hr025Cl; HSPC192; ILFS1; KIAA1352; LARS; LARS1; LEURS; LEUS; LFIS; LRS; mKIAA1352; PIG44; RNTLS
UniProt ID: (Human) Q9P2J5, (Mouse) Q8BMJ2
Entrez Gene ID: (Human) 51520, (Mouse) 107045
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support