Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSWESGAGP GLGSQGMDLV WSAWYGKCVK GKGSLPLSAH GIVVAWLSRA EWDQVTVYLF CDDHKLQRYA LNRITVWRSR SGNELPLAVA STADLIRCKL LDVTGGLGTD ELRLLYGMAL VRFVNLISER KTKFAKVPLK CLAQEVNIPD WIVDLRHELT HKKMPHINDC RRGCYFVLDW LQKTYWCRQL ENSLRETWEL EEFREGIEEE DQEEDKNIVV DDITEQKPEP QDDGKSTESD VKADGDSKGS EEVDSHCKKA LSHKELYERA RELLVSYEEE QFTVLEKFRY LPKAIKAWNN PSPRVECVLA ELKGVTCENR EAVLDAFLDD GFLVPTFEQL AALQIEYEDG QTEV (1-353 aa encoded by BC014545)
Involved in the biogenesis of the 60S ribosomal subunit. Required for maturation of the 28S rRNA. Functions as a component of the Five Friends of Methylated CHTOP (5FMC) complex; the 5FMC complex is recruited to ZNF148 by methylated CHTOP, leading to desumoylation of ZNF148 and subsequent transactivation of ZNF148 target genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endoribonuclease LAS1L; LAS1-like, ribosome biogenesis factor; Protein LAS1 homolog; Ribosomal biogenesis protein LAS1L
Gene Aliases: 1810030A06Rik; 5830482G23Rik; dJ475B7.2; Las1-like; LAS1L; MSTP060; RGD1565755
UniProt ID: (Human) Q9Y4W2, (Mouse) A2BE28
Entrez Gene ID: (Human) 81887, (Mouse) 76130, (Rat) 296865
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support