The recommended shelf life for this product is 1 year from date of receipt.
Involved in the biogenesis of the 60S ribosomal subunit. Required for maturation of the 28S rRNA. Functions as a component of the Five Friends of Methylated CHTOP (5FMC) complex; the 5FMC complex is recruited to ZNF148 by methylated CHTOP, leading to desumoylation of ZNF148 and subsequent transactivation of ZNF148 target genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endoribonuclease LAS1L; LAS1-like, ribosome biogenesis factor; Protein LAS1 homolog; Ribosomal biogenesis protein LAS1L
Gene Aliases: 1810030A06Rik; 5830482G23Rik; dJ475B7.2; Las1-like; LAS1L; MSTP060
UniProt ID: (Human) Q9Y4W2, (Mouse) A2BE28
Entrez Gene ID: (Human) 81887, (Mouse) 76130
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support