Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EFEKSRMGPS GGEGMEPERR DSQDGSSYRR PLEQQQPHHI PTSAPVYQQP QQQPVAQSYG GYKEPAAPVS IQRSAPGGGG KRYRAA
This gene encodes a member of a LIM protein subfamily characterized by a LIM motif and a domain of Src homology region 3. The encoded protein functions as an actin-binding protein and possibly in cytoskeletal organization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LASP-1; LIM and SH3 domain protein 1; Metastatic lymph node gene 50 protein; MLN 50
Gene Aliases: Lasp-1; LASP1; MLN50
UniProt ID: (Human) Q14847
Entrez Gene ID: (Human) 3927
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support