Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: ERFIGLPLSR WLGVRDQTRR QVKPNATLEK HFLTEGHRPK EPQLSLLAAQ CGLTLQQTQR WFRRRRNQDR PQLTKKFCEA SWR
The LASS (longevity assurance homolog) family members are highly conserved from yeasts to mammals. Six members of this family of proteins have been characterized (LASS1, LASS2, LASS3, LASS4, LASS5 and LASS6) and they are all involved in sphingolipid synthesis. LASS4 is a 394 amino acid endoplasmic reticulum, multi-pass membrane protein. LASS4 increases the levels of long ceramides such as C22:0- and C24:0-ceramides. In cells deficient for CLN9, as observed in neuronal ceroid lipofuscinosis (NCL) or Batten disease, LASS4 can increase ceramide levels and partially correct growth and apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ceramide synthase 4; CerS4; FLJ12089; LAG1 homolog, ceramide synthase 4; LAG1 longevity assurance homolog 4; Sphingosine N-acyltransferase CERS4
Gene Aliases: CERS4; LASS4; Trh1
UniProt ID: (Human) Q9HA82
Entrez Gene ID: (Human) 79603
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support