Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEEAILVPC VLGLLLLPIL AMLMALCVHC HRLPGSYDST SSDSLYPRGI QFKRPHTVAP WPPAYPPVTS YPPLSQPDLL PIPRSPQPLG GSHRTPSSRR DSDGANSVAS YENEEPACED ADEDEDDYHN PGYLVVLPDS TPATSTAAPS APALSTPGIR DSAFSMESID DYVNVPESGE SAEASLDGSR EYVNVSQELH PGAAKTEPAA LSSQEAEEVE EEGAPDYENL QELN (1-262 aa encoded by BC011563)
LAT (Linker for Activation of T cells) is a 36-38 kDa double-palmitoylated transmembrane adaptor protein expressed by T cells, pre-B cells, NK cells, mast cells and platelets. After immunoreceptor triggering, LAT becomes multiply tyrosine-phosphorylated by Syk-, Src-, or Tec-family kinases, providing docking sites for downstream signaling molecules. LAT is essential for TCR-dependent T cell- and FcepsilonRI-dependent mast cell activation, as well as for maturation of early thymocytes. It is also involved in NK cell signaling and platelet activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 36 kDa phospho-tyrosine adapter protein; 36 kDa phospho-tyrosine adaptor protein; 36 kDa phosphotyrosine adapter protein; linker for activation of T cells; linker for activation of T cells, transmembrane adaptor; Linker for activation of T-cells family member 1; linker protein; p36-38; pp36
Gene Aliases: LAT; LAT1; pp36
UniProt ID: (Human) O43561, (Mouse) O54957
Entrez Gene ID: (Human) 27040, (Mouse) 16797
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support