Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Negatively regulates TCR (T-cell antigen receptor)-mediated signaling in T-cells and BCR (B-cell antigen receptor)-mediated signaling in B-cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LAT-like membrane associated protein; Linker for activation of X cells; Lymphocyte transmembrane adapter 1; Membrane-associated adapter protein LAX
Gene Aliases: LAX; LAX1
UniProt ID: (Human) Q8IWV1
Entrez Gene ID: (Human) 54900
Molecular Function:
defense/immunity protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support