Immunogen sequence: TNHQIYELEK RFLYQKYLSP ADRDQIAQQL GLTNAQVITW FQNRRAKLKR DLEEMKADVE SAKKLGPSGQ MDIVALAELE QNSEATA
LBX1 is the transcription factor required for the development of GABAergic interneurons in the dorsal horn of the spinal cord and migration and further development of hypaxial muscle precursor cells for limb muscles, diaphragm and hypoglossal cord.This gene and the orthologous mouse gene were found by their homology to the Drosophila lady bird early and late homeobox genes. In the mouse, this gene is a key regulator of muscle precursor cell migration and is required for the acquisition of dorsal identities of forelimb muscles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lady bird-like homeobox; ladybird homeobox homolog 1; Ladybird homeobox protein homolog 1; Transcription factor LBX1; transcription factor similar to D. melanogaster homeodomain protein lady bird late
Gene Aliases: homeobox; HPX-6; HPX6; LBX1; LBX1H
UniProt ID: (Human) P52954
Entrez Gene ID: (Human) 10660
Molecular Function:
helix-turn-helix transcription factor
homeodomain transcription factor
DNA-binding transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support