Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: ELERRFVFQK YLAPSERDGL ATRLGLANAQ VVTWFQNRRA KLKRDVEEMR ADVASLRALS PEVLCSLALP EGAPDPGLCL GPAGPDSRPH LSDEEIQVDD
Putative transcription factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lady bird-like homeobox 2; Ladybird homeobox 2; ladybird homeobox homolog 2; Ladybird homeobox protein homolog 2; ladybird-like homeobox 2; ladybird-like homeodomain protein 2; Transcription factor LBX2
Gene Aliases: LBX2; LP3727
UniProt ID: (Human) Q6XYB7
Entrez Gene ID: (Human) 85474
Molecular Function:
helix-turn-helix transcription factor
homeodomain transcription factor
DNA-binding transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support