Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MAP1LC3A (Microtubule Associated Protein 1 Light Chain 3 Alpha) is a Protein Coding gene. Diseases associated with MAP1LC3A include Machado-Joseph Disease and Neurodegeneration With Brain Iron Accumulation. Among its related pathways are Selective autophagy and Mitophagy. Gene Ontology (GO) annotations related to this gene include microtubule binding and phosphatidylethanolamine binding. An important paralog of this gene is MAP1LC3B. MAP1LC3B (Microtubule Associated Protein 1 Light Chain 3 Beta) is a Protein Coding gene. Diseases associated with MAP1LC3B include Liver Carcinoma In Situ and Pheochromocytoma. Among its related pathways are Early SARS-CoV-2 Infection Events and Selective autophagy. Gene Ontology (GO) annotations related to this gene include microtubule binding. MAP1LC3C (Microtubule Associated Protein 1 Light Chain 3 Gamma) is a Protein Coding gene. Diseases associated with MAP1LC3C include Machado-Joseph Disease. Among its related pathways are Selective autophagy and Ferroptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Autophagy-related protein LC3 A; Autophagy-related protein LC3 B; Autophagy-related protein LC3 C; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related ubiquitin-like modifier LC3 C; LC3-like protein 2; MAP1 light chain 3-like protein 1; MAP1 light chain 3-like protein 2; MAP1 light chain 3-like protein 3; MAP1A/1B light chain 3 A; MAP1A/1B light chain 3 B; MAP1A/MAP1B LC3 A; MAP1A/MAP1B LC3 B; MAP1A/MAP1B LC3 C; MAP1A/MAP1B light chain 3 A; MAP1A/MAP1B light chain 3 B; MAP1A/MAP1B light chain 3 C; Microtubule-associated protein 1 light chain 3 alpha; Microtubule-associated protein 1 light chain 3 beta; Microtubule-associated protein 1 light chain 3 gamma; microtubule-associated proteins 1A/1B light chain 3; Microtubule-associated proteins 1A/1B light chain 3A; Microtubule-associated proteins 1A/1B light chain 3B; Microtubule-associated proteins 1A/1B light chain 3C
Gene Aliases: 1010001C15Rik; 1010001H21Rik; 4922501H04Rik; Atg8; ATG8E; ATG8F; ATG8J; LC3; LC3A; LC3B; LC3C; MAP1A/1BLC3; MAP1ALC3; MAP1BLC3; Map1lc3; MAP1LC3A; MAP1LC3B; MAP1LC3B-a; MAP1LC3C; Mpl3; zbs559
UniProt ID: (Human) Q9H492, (Human) Q9GZQ8, (Human) Q9BXW4, (Rat) Q6XVN8, (Mouse) Q91VR7, (Rat) Q62625, (Mouse) Q9CQV6
Entrez Gene ID: (Human) 84557, (Human) 81631, (Human) 440738, (Rat) 362245, (Mouse) 66734, (Rat) 64862, (Mouse) 67443
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support