Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LCoR; Ligand-dependent corepressor; Mblk1-related protein 2
Gene Aliases: C10orf12; KIAA1795; LCOR; MLR2
UniProt ID: (Human) Q96JN0
Entrez Gene ID: (Human) 84458
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support