Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GPSIVCKSIQ MNQAENSLQE EQEGPLDLTV NRMQEQNTQQ GDGVLDLSTK KTSIKSEESS ICDPSSENSV AGRLHRNRED YVERSAEFAD GLL
Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 65%.
May act as transcription activator that binds DNA elements with the sequence 5'-CCCTATCGATCGATCTCTACCT-3'. May play a role in spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LCOR-like protein; Ligand-dependent nuclear receptor corepressor-like protein; MBLK1-related protein; transcription factor MLR1
Gene Aliases: LCORL; MLR1
UniProt ID: (Human) Q8N3X6
Entrez Gene ID: (Human) 254251
Molecular Function:
DNA-binding transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support