Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: AGPLTNDLLH NLSGLLGDQS SNFVAGDKDM YVCHQPLPTF LPEYFSSLHA SQITHYKVFL SWAQLLPAGS TQNPDEKTVQ CYRRLLKALK TARLQPMV
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lactase-glycosylceramidase; lactase-phlorizin hydrolase-1; Lactase/glycosylceramidase; Lactase/phlorizin hydrolase
Gene Aliases: LAC; LCT; LPH; LPH1
UniProt ID: (Human) P09848
Entrez Gene ID: (Human) 3938
Molecular Function:
glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support