Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody reacts with isoform 1.
Immunogen sequence: MLDRDVGPT PMYPPTYLEP GIGRHTPYGN QTDYRIFELN KRLQNWTEEC DNLWWDAFTT EFFEDDAMLT ITFCLEDGPK RYTIGRTLIP RYFRSIFEGG ATELYYVLKH PKEAFHSNFV SLDCDQGSMV TQHGKPMFTQ VCVEGRLYLE FMFDDMMRIK TWHFSIRQHR ELIPRSILAM HAQDPQMLDQ LSKNITRCGL SNSTLNYLRL CVI (1-375 aa encoded by BC000482)
LIM domain-binding protein 1 (LDB1), also known as Carboxyl-terminal LIM domain-binding protein 2 (CLIM2), binds to the LIM domain of a wide variety of LIM domain-containing transcription factors. LDB1 may regulate the transcriptional activity of LIM-containing proteins by determining specific partner interactions. It plays a role in the development of interneurons and motor neurons in cooperation with LHX3 and ISL1. LDB1 acts synergistically with LHX1/LIM1 in axis formation and activation of gene expression. It also acts with LMO2 in the regulation of red blood cell development, maintaining erythroid precursors in an immature state. LDB1 strongly interacts with the LIM2 domain of LMX1A to form homodimer. LDB1 protein is expressed in a wide range of adult tissues including brain, heart, skeletal muscle, colon, thymus, spleen, kidney, liver, small intestine, lung and peripheral blood leukocytes. Due to alternative splicing events, three isoforms exist for LDB1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carboxy terminal LIM domain protein 2; Carboxyl-terminal LIM domain-binding protein 2; CLIM-2; hLdb1; LDB-1; LIM domain-binding factor CLIM2; LIM domain-binding factor-1; LIM domain-binding protein 1; Nuclear LIM interactor
Gene Aliases: CLIM-2; CLIM2; LDB-1; LDB1; NLI
UniProt ID: (Human) O75479
Entrez Gene ID: (Human) 8861
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support