Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MATLKEKLI APVAEEEATV PNNKITVVGV GQVGMACAIS ILGKSLADEL ALVDVLEDKL KGEMMDLQHG SLFLQTPKIV ADKDYSVTAN SKIVVVTAGV RQQEGESRLN LVQRNVNVFK FIIPQIVKYS PDCIIIVVSN PVDILTYVTW KLSGLPKHRV IGSGCNLDSA RFRYLMAEKL GIHPSSCHGW ILGEHGDSSV AVWSGVNVAG VSLQELNPEM GTDNDSENWK EVHKMVVESA YEVIKLKGYT NWAIGLSVAD LIESMLKNLS RIHPVSTMVK GMYGIENEVF LSLPCILNAR GLTSVINQKL KDDEVAQLKK SADTLWDIQK DLKDL (1-334 aa encoded by BC002362)
This gene encodes an enzyme which catalyzes the reversible conversion of lactate and pyruvate, and NAD and NADH, in the glycolytic pathway. Mutations in this gene are associated with lactate dehydrogenase B deficiency. Pseudogenes have been identified on the X chromosome and on chromosome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 281; L-lactate dehydrogenase B chain; lactate dehydrogenase H chain; Lactate dehydrogenease B; LDH heart subunit; LDH-B; LDH-H; LDHH; Renal carcinoma antigen NY-REN-46; testicular secretory protein Li 25
Gene Aliases: AI790582; H-Ldh; HEL-S-281; Ldh-2; LDH-B; LDH-H; Ldh2; LDHB; LDHBD; TRG-5
UniProt ID: (Human) P07195, (Rat) P42123, (Mouse) P16125
Entrez Gene ID: (Human) 3945, (Rat) 24534, (Mouse) 16832
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support