Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody can recognize all 3 isoforms of LDLRAD1.
LDLRAD1 belongs to the LDLR family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Low-density lipoprotein receptor class A domain-containing protein 1
Gene Aliases: LDLRAD1; OTTMUSG00000008594
UniProt ID: (Human) Q5T700
Entrez Gene ID: (Human) 388633, (Mouse) 546840
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support