Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total LDOC1L.
LDOC1L gene ontology (GO) annotation includes protein binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6-Mar; gag-like protein; Leucine zipper protein down-regulated in cancer cells-like; leucine zipper, down-regulated in cancer 1 like; leucine zipper, down-regulated in cancer 1-like; Mammalian retrotransposon-derived protein 6; Protein LDOC1L; Protein Sushi-15E3; Retrotransposon Gag-like protein 6
Gene Aliases: BC058638; dJ1033E15.2; LDOC1L; MAR6; MART6; RTL6; Sushi-15E3
UniProt ID: (Human) Q6ICC9, (Mouse) Q505G4
Entrez Gene ID: (Human) 84247, (Mouse) 223732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support