Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RFSQSFREVA GRFLASEAST HLLVFGMEQR LPPNSELVQA VLRLFQEPVP KAALHRHGRL SPRSAQARVT VEWLRVRDDG SNRTSLIDSR LVSVHESGWK AFDV
Left-right determination factor 1 and 2 (Lefty1/2) belong to the TGF-beta superfamily of proteins and are responsible for determination of left-right positioning of visceral organs during embryogenesis. Lefty1 acts as a regulator of Lefty2 and NODAL as the lack of the former resulted in bilateral expression of nodal, lefty2 and Pitx2 (a homeobox gene normally expressed on the left side and blocks the signal for leftness). Lefty2 may play a role in endometrial bleeding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endometrial bleeding-associated factor; Left-right determination factor 2; Left-right determination factor A; MGC46222; Protein lefty-2; Protein lefty-A; TGF-beta-4; Transforming growth factor beta-4
Gene Aliases: EBAF; LEFTA; LEFTY2; LEFTYA; PSEC0024; TGFB4
UniProt ID: (Human) O00292
Entrez Gene ID: (Human) 7044
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support