Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: TPTPFTNAVQ LLLTLKLVSL ASEVQDLHLA QRKEMASGFS KGPTLGLLPD VPSLMETLSY SYCYVGIMTG PFFRYRTYLD WLEQPFPGAV PSLRPLL
MBOAT7 belongs to the membrane-bound acyltransferase family. It is involved in arachidonate recycling, thus regulating free arachidonic acid levels and leukotriene synthesis in neutrophils.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acylglycerophosphatidylinositol O-acyltransferase; Bladder and breast carcinoma-overexpressed gene 1 protein; h-mboa-7; leukocyte receptor cluster (LRC) member 4; Leukocyte receptor cluster member 4; LPIAT; LPLAT 7; lyso-PI acyltransferase; Lysophosphatidylinositol acyltransferase; Lysophospholipid acyltransferase 7; malignant cell expression-enhanced gene/tumor progression-enhanced; Membrane-bound O-acyltransferase domain-containing protein 7; O-acyltransferase domain-containing protein 7
Gene Aliases: BB1; hMBOA-7; LENG4; LPIAT; LRC4; MBOA7; MBOAT7; OACT7
UniProt ID: (Human) Q96N66
Entrez Gene ID: (Human) 79143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support