Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Negatively regulates leptin receptor (LEPR) cell surface expression, and thus decreases response to leptin. Negatively regulates growth hormone (GH) receptor cell surface expression in liver. May play a role in liver resistance to GH during periods of reduced nutrient availability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily C, member 6; Endospanin-1; leptin receptor gene related protein; Leptin receptor gene-related protein; Leptin receptor overlapping transcript protein; OB-R gene-related protein; OB-RGRP
Gene Aliases: LEPR; LEPROT; OB-RGRP; OBR; OBRGRP; VPS55
UniProt ID: (Human) O15243, (Pig) B9TRX0
Entrez Gene ID: (Human) 54741, (Pig) 100270821
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support