Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This gene encodes a protein that is localized to the inner mitochondrial membrane. The protein functions to maintain the mitochondrial tubular shapes and is required for normal mitochondrial morphology and cellular viability. Mutations in this gene cause Wolf-Hirschhorn syndrome, a complex malformation syndrome caused by the deletion of parts of the distal short arm of chromosome 4. Related pseudogenes have been identified on chromosomes 8, 15 and 19.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Electroneutral mitochondrial K(+)/H(+)exchanger; KHE; LETM1 and EF-hand domain-containing protein 1, mitochondrial; leucine zipper-EF-hand containing transmembrane protein 1; Leucine zipper-EF-hand-containing transmembrane protein 1; Mdm38 homolog; Mitochondrial proton/calcium exchanger protein
Gene Aliases: LETM1
UniProt ID: (Human) O95202, (Rat) Q5XIN6, (Mouse) Q9Z2I0
Entrez Gene ID: (Human) 3954, (Rat) 305457, (Mouse) 56384
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support