Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSQPSGGRAP GTRIYSWSCP TVMSPGEKLD PIPDSFILQP PVFHPVVPYV TTIFGGLHAG KMVMLQGVVP LDAHRFQVDF QCGCSLCPRP DIAFHFNPRF HTTKPHVICN TLHGGRWQRE ARWPHLALRR GSSFLILFLF GNEEVKVSVN GQHFLHFRYR LPLSHVDTLG IFGDILVEAV GFLNINPFVE GSREYPAGHP FLLMSPRLEV PCSHALPQGL SPGQVIIVRG LVLQEPKHFT VSLRDQAAHA PVTLRASFAD RTLAWISRWG QKKLISAPFL FYPQRFFEVL LLFQEGGLKL ALNGQGLGAT SMNQQALEQL RELRISGSVQ LYCVHS
Galectin-12, also designated galectin-related inhibitor of proliferation 1 in mouse, is a 314 amino acid protein encoded by the human gene LGALS12. Galectin-12 is a member of the galectin family consisting of b-galactoside-binding proteins with conserved carbohydrate recognition domains. Galectin-12 binds lactose and may participate in the apoptosis of adipocytes. This protein is preferentially expressed in peripheral blood leukocytes and adipocytes. Galectin-12 is induced by cell cycle block at the G1 phase and causes G1 arrest when overexpressed. The galectin-12 gene is expressed in mouse preadipo-cytes and is upregulated when preadipocytes undergo cell cycle arrest, concomitant with acquisition of the competence to undergo differentiation in response to adipogenic hormone stimulation. Galectin-12 is an adipocyte-expressed protein which is downregulated by various insulin resistance-inducing hormones. As a result, galectin-12 may play a role in the pathogenesis of insulin resistance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gal-12; Galectin-12; Galectin-related inhibitor of proliferation; lectin, galactoside binding soluble 12; lectin, galactoside-binding, soluble, 12; testicular secretory protein Li 26
Gene Aliases: GAL12; GRIP1; LGALS12
UniProt ID: (Human) Q96DT0
Entrez Gene ID: (Human) 85329
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support