Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HQALHPWHR DTDLEFVDGE GKPRLIVSSS SQAPVIYQWS RTQKQFVAQG EVTQVPDAQA VKHFRAGRDS YLCLSRYIGD SKILRWEGTR FSEVQALPSR GSLALQPFLV GGRRYLALGS DFSFTQIYQW DEGRQKFVRF QELAVQAPRA FCYMPAGDAQ LLLAPSFKGQ TLVYRHIVVD LSA (367-548 aa encoded by BC142997)
The leucine-rich, glioma inactivated gene 3 (LGI3) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI3 have not been found to be associated with ADTLE. LGI3 expression is induced in rat astrocyte cultures by the amyloid beta (Abeta) peptide and accumulated on neuronal plasma membranes of aged monkey brains and co-localized with Abeta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leubrin; Leucine-rich glioma-inactivated protein 3; Leucine-rich repeat LGI family member 3; leucine-rich repeat LGI family, member 3; LGI1-like protein 4
Gene Aliases: AI841179; AW049851; LGI3; LGIL4; UNQ8190/PRO23199
UniProt ID: (Human) Q8N145, (Mouse) Q8K406
Entrez Gene ID: (Human) 203190, (Rat) 306013, (Mouse) 213469
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support