Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PLCAAPCSC DGDRRVDCSG KGLTAVPEGL SAFTQALQLA GNDLSFIHPK ALSGLKELKV LTLQNNQLKT VPSEAIRGLS ALQSLRLDAN HITSVPEDSF EGLVQLRHLW LDDNSLTEVP VHHLSNLPTL QALTLALNKI SSIPDFAFTN LSSLVVLHLH NNKIRSLSQH CFDGLDNLET LDLNYNNLGE FPQAIKALPS LKELGFHSNS ISVIPDGAFD GNPLLRTIHL YDNPLSFVGN SAFHNLSDLH SLVIRGASMV QQFPNLTGTV HLESLTLTGT KISSIPNNLC QEQKMLRTLD LSYNNIRDLP SFNGCHALEE ISLQRNQIYQ IKEGTFQGLI SLRILDLSRN L (27-376 aa encoded by BC033039)
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR LGR4 is a member of this family (subfamily Orphan-A). Its expression has been documented in human brain, adrenal, heart, kidney, liver, skeletal muscle, ovary, pancreas, placenta, prostate, spleen, stomach, testis, thyroid, and spinal cord.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 48; G protein-coupled receptor GPR48; G-protein coupled receptor 48; leucine-rich repeat containing G protein-coupled receptor 4; Leucine-rich repeat-containing G-protein coupled receptor 4
Gene Aliases: 9130225G07; A330106J01Rik; A930009A08Rik; BNMD17; GPR48; LGR4
UniProt ID: (Human) Q9BXB1, (Mouse) Q80T31, (Rat) Q9Z2H4
Entrez Gene ID: (Human) 55366, (Mouse) 107515, (Rat) 286994
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support