Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FPVTPEAVKS VLLVVLPLPA CLNPLLYLLF NPHFRDDLRR LRPRAGDSGP LAYAAAGELE KSSCDSTQAL VAFSDVDLIL EASEAGRPPG LETYGFPSVT
LGR6 has been reported to be expressed in rat adrenal, brain, colon, heart, kidney, ovary, oviduct, small intestine, spleen, testis, and uterus. ESTs have been isolated from human B-cell/lung/testis, blood, brain, colon, eye, germ cell, head/neck, heart, heart/melanocyte/uterus, lung, ovary, stomach, testis, and uterus libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ14471; gonadotropin receptor; leucine-rich repeat containing G protein-coupled receptor 6; leucine-rich repeat-containing G protein-coupled receptor 6; Leucine-rich repeat-containing G-protein coupled receptor 6; RP11-572A16.3
Gene Aliases: GPCR; LGR6; RGD1560339; UNQ6427/PRO21331; VTS20631
UniProt ID: (Human) Q9HBX8
Entrez Gene ID: (Human) 59352, (Rat) 498233
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support