Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: CTDRSLSPDL QDALQDDPKE TDNSTSSDKE TANNENEEQN SGTKRRGPRT TIKAKQLETL KAAFAATPKP TRHIREQLAQ ETGLNMRVIQ VWFQNRRSKE
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in the control of differentiation and development of the forebrain. In mice, this protein is essential for the regulation of precursor cell proliferation and the control of neuronal differentiation and migration during hippocampal development. This protein is involved in learning and motor functions in adult mice.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LIM homeobox protein 5; LIM/homeobox protein Lhx5; MGC129689
Gene Aliases: LHX5
UniProt ID: (Human) Q9H2C1
Entrez Gene ID: (Human) 64211
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support