Peptide Sequence: HKKHTPQHPV PPSGAPPSRL PSALSDDIHY TPFSSPERAR MVTLHGYIES QVQCGQVHCR LPYTAPPVHL KADMDGPLSN RGEKVILFQY
LHX6 (LIM/homeobox protein Lhx6) is a probably transcription factor required for the expression of a subset of genes involved in interneurons migration and development. It functions in the specification of cortical interneuron subtypes and in the migration of GABAergic interneuron prescursors from the subpallium to the cerebral cortex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LIM homeobox protein 6; LIM homeodomain protein 6.1; LIM/homeobox protein Lhx6; LIM/homeobox protein Lhx6.1; MGC119542; MGC119544; MGC119545; RP11-498E2.6
Gene Aliases: LHX6; LHX6.1
UniProt ID: (Human) Q9UPM6
Entrez Gene ID: (Human) 26468
Molecular Function:
helix-turn-helix transcription factor
homeodomain transcription factor
DNA-binding transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support