Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSLRCGDAAR TLGPRVFGRY FCSPVRPLSS LPDKKKELLQ NGPDLQDFVS GDLADRSTWD EYKGNLKRQK GERLRLPPWL KTEIPMGKNY NKLKNTLRNL NLHTVCEEAR CPNIGECWGG GEYATATATI MLMGDTCTRG CRFCSVKTAR NPPPLDASEP YNTAKAIAEW GLDYVVLTSV DRDDMPDGGA EHIAKTVSYL KERNPKILVE CLTPDFRGDL KAIEKVALSG LDVYAHNVET VPELQSKVRD PRVNFDQSLR VLKHAKKVQP DVISKTSIML GLGENDEQVY ATMKALREAD VDCLTLGQYM QPTRRHLKVE EYITPEKFKY WEKVGNELGF HYTASGPLVR SSYKAGEFFL KNLVAKRKTK DL
LIAS belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LIAS; lip-syn; lip-syn {ECO:0000255|HAMAP-Rule:MF_03123}; Lipoate synthase; lipoate synthase {ECO:0000255|HAMAP-Rule:MF_03123}; Lipoic acid synthase; lipoic acid synthase {ECO:0000255|HAMAP-Rule:MF_03123}; Lipoyl synthase, mitochondrial; lipoyl synthase, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03123}; LS; LS {ECO:0000255|HAMAP-Rule:MF_03123}; MGC23245
Gene Aliases: 2900022L22Rik; 4933425M12Rik; 7a5ex; C77512; HGCLAS; HUSSY-01; LAS; LIAS; LIP1; LS; mLip1; PDHLD
UniProt ID: (Human) O43766, (Rat) Q5XIH4, (Mouse) Q99M04
Entrez Gene ID: (Human) 11019, (Pig) 100627627, (Rat) 305348, (Mouse) 79464
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support