Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blots, this antibody shows less than 10% cross-reactivity with recombinant mouse (rm) EDA, rmTRAIL, rmTRANCE, rmTWEAK, and rhVEGI but does not cross-react rm4-1BB Ligand, rhAPRIL, rmBAFF, rhEDA-A2, rmFas Ligand, rhGITR Ligand, rmGITR Ligand, rhLymphotoxin alpha 1/ beta 2, rhLymphotoxin alpha 2/ beta 2, rmOX40 Ligand, and rmTNF-alpha.
Reconstitute at 0.5 mg/mL in sterile PBS.
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD258; sCD258; sLIGHT; soluble CD258; soluble LIGHT; Tumor necrosis factor ligand superfamily member 14
Gene Aliases: HVEM-L; HVEML; LIGHT; LTg; Ly113; Tnfsf14
UniProt ID: (Mouse) Q9QYH9
Entrez Gene ID: (Mouse) 50930
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support