Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody has been tested in indirect ELISA.
LILRA6 may act as receptor for class I MHC antigens.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: immunoglobulin-like transcript 5; immunoglobulin-like transcript 8; leucocyte Ig-like receptor A6; leukocyte Ig-like receptor; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 6; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 6
Gene Aliases: CD85b; ILT-8; ILT5; ILT8; LILRB3; LILRB6
Entrez Gene ID: (Human) 79168
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support