Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MKMRFLGLVV CLVLWPLHSE GSGGKLTALD PETNMNVSEI ISYWGFPSEE YLVETEDGYI LCLNRIPHGR KNHSDKGPKP VVFLQHGLLA DSSNWVTNLA NSSLGFILAD AGFDVWMGNS RGNTWSRKHK TLSVSQDEFW AFSYDEMAKY DLPASINFIL NKTGQEQVYY VGHSQGTTIG FIAFSQIPEL AKRIKMFFAL GPVASVAFCT SPMAKLGRLP DHLIKDLFGD KEFLPQSAFL KWLGTHVCTH VILKELCGNL CFLLCGFNER NLNMSRVDVY TTHSPAGTSV QNMLHWSQAV KFQKFQAFDW GSSAKNYFHY NQSYPPTYNV KDMLVPTAVW SGGHDWLADV YDVNILLTQI TNLVFHESIP EWEHLDFIWG LDAPWRLYNK IINLMRKYQ
This gene encodes lipase A, the lysosomal acid lipase (also known as cholesterol ester hydrolase). This enzyme functions in the lysosome to catalyze the hydrolysis of cholesteryl esters and triglycerides. Mutations in this gene can result in Wolman disease and cholesteryl ester storage disease. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid cholesteryl ester hydrolase; cholesterol ester hydrolase; Cholesteryl esterase; Lipase A; lipase A, lysosomal acid, cholesterol esterase; lysosomal acid lipase; Lysosomal acid lipase/cholesteryl ester hydrolase; RP11-341B24.1; Sterol esterase
Gene Aliases: CESD; LAL; LIPA
UniProt ID: (Human) P38571
Entrez Gene ID: (Human) 3988
Molecular Function:
hydrolase
lipase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support